missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Smooth Muscle Actin Mouse, Clone: 1A4/asm-1, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 23 publications
Marca: Bio-Techne NBP2-33006-0.2MG
679.20 EUR valido fino al 2025-03-28
Usa il codice promo "25306" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Baboon, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifica
alpha-Smooth Muscle Actin | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 0.5-1ug/ml, Simple Western 10 ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 1:600 | |
P62736 | |
ACTA2 | |
N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH | |
Protein A purified | |
RUO | |
59 | |
Human, Mouse, Rat, Porcine, Baboon, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Monkey, Rabbit, Sheep | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
1A4/asm-1 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
1a4 sma, 1a4 smooth muscle actin, AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTVS, alpha 2 actin, alpha SMA, alpha sma 1a4, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, sma 1a4, smooth muscle actin | |
Mouse | |
42 kDa | |
0.2 mg | |
Primary | |
Actin is a major component of the cytoskeleton and is present in most cell types. This MAb is highly specific to actin from smooth muscles. Its epitope lies in the first four N-terminal amino acids. This MAb does not stain cardiac or skeletal muscle; however, it does stain myofibroblasts and myoepithelial cells. This antibody could be used together with anti-muscle specific actin and myogenin in making a diagnosis of smooth muscle and skeletal muscle tumors. In most cases of rhabdomyosarcoma, this antibody yields negative results whereas anti-muscle specific actin and myogenin are positive. Leiomyosarcomas are positive only with anti-muscle specific actin and anti-smooth muscle actin and are negative with anti-myogenin. | |
Store at 4C. | |
IgG2a κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
alpha-Smooth Muscle Actin Mouse, Clone: 1A4/asm-1, Novus Biologicals™ > 0.2mg; Unlabeled
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto