Learn More
Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets
Provides exceptional handling characteristics, allowing reprobing
Marca: Cytiva 10600054
Dettagli aggiuntivi : Peso : 0.15500kg
Descrizione
Individual sheets.
- Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
- The support does not affect transfer conditions or results
Specifica
20 cm | |
10/Pk. | |
Nitrocellulose, Inert Polyester |
20 cm | |
Sheet | |
0.2 μm |