Learn More
Laboratoires Anios™ Aniosafe Savon Doux HF
Laboratoires Anios™ Aniosafe Savon Doux HF is a gentle soap recommended for frequent hand hygiene and general body cleansing in hospital environments.
Marca: Laboratoires Anios™ 1918228VB
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Dermatologically tested
- Enriched with moisturizer
- Preservation of the product demonstrated according to NF EN ISO 11930
Specifica
Gentle Soap | |
Water, Disodium Lauroamphodiacetate, Sodium Lauryl Sulfate, Hexylene Glycol, Glycerin, Linoleamide DEA, Disodium Ricinoleamido EASulfosuccinate, Benzyl Alcohol, Sodium Benzoate, PEG-200 Hydrogenated Glyceryl Palmate and PEG-7 Glyceryl Cocoate, Parfum (Fragrance), Lactic acid. | |
Bottles with screw pump | |
500 mL |
NF EN ISO 11930 | |
General body cleansing in hospital environments | |
12 Bottles |
Titolo del prodotto
Laboratoires Anios™ Aniosafe Savon Doux HF > 12 x 500mL bottles with screwed pump