missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16/CD32 Rat anti-Mouse, Unlabeled, Clone: 2.4G2, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 553141
348.82 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The 2.4G2 antibody reacts specifically with a common nonpolymorphic epitope on the extracellular domains of the mouse Fc γ III and Fc γ II receptors. It has also been reported to bind the Fc γ I receptor (CD64) via its Fc domain. 2.4G2 mAb blocks non-antigen-specific binding of immunoglobulins to the Fc γ III and Fc γ II, and possibly Fc γ I, receptors in vitro and in vivo . CD16 and/or CD32 are expressed on natural killer cells, monocytes, macrophages, dendritic cells (at low levels), Kupffer cells, granulocytes, mast cells, B lymphocytes, immature thymocytes, and some activated mature T lymphocytes.
Specifica
CD16/CD32 | |
Monoclonal | |
0.5mg/mL | |
FcγRIII/FcγRII; Fcgr3/Fcgr2 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Blocking Assay, Flow Cytometry | |
2.4G2 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse BALB/c Macrophage J774 Cell Line | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
CD16/CD32 Rat anti-Mouse, Unlabeled, Clone: 2.4G2, BD > 0.1mg; Unlabeled
For Research Use Only.
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto