Learn More
BCLAF1 Mouse, anti-Human, Unconjugated, Clone: 1D6G7, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 67860-1-IG-150UL
Codice prodotto. 17281526
  • 399.00€ / 150µL
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
BCLF1, also named as Bcl-2-associated transcription factor 1, is a 920 amino acid protein, which Interacts with Bcl-2 related proteins, EMD, with the adenovirus E1B 19 kDa protein and with DNA. BCLF1 as a death-promoting transcriptional repressor may be involved in cyclin-D1/CCND1 mRNA stability through the SNARP complex which associates with both the 3′end of the CCND1 gene and its mRNA. The calculated molecular weight of BCLF1 is a 110 kDa, but the modified BCLF1 protein is about 140-150 kDa.
Specifica
Specifica
BCLAF1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunofluorescence, ELISA
1D6G7
Unconjugated
150 μL
9774