Learn More
BD BBL™ Mycoflask™ Sabouraud Dextrose Agar Prepared Media Slants
Used in qualitative procedures for cultivation of dermatophytes
Marca: BD 221013
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifica
CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP | |
2°C to 8°C | |
United States | |
Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L | |
Tube with Tight Cap |
Beige | |
Liquid | |
Sabouraud Dextrose Agar | |
Cultivation of dermatophytes | |
100 Pack |
For in vitro diagnostic use.