missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Selenite-F Broth Prepared Media
For isolation of Salmonella from feces, urine, water, foods and other materials of sanitary importance
Marca: BD 221020
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifica
CE, BS12, MCM9, SMWW | |
2°C to 8°C | |
United States | |
Pancreatic digest of Casein 5g/L, Lactose 4g/L, Sodium Selenite 4g/L, Sodium Phosphate 10g/L | |
Tube with Tight Cap |
Beige | |
Liquid | |
Selenite-F Broth | |
Isolation of Salmonella from feces, urine, Water, foods and other materials of sanitary importance | |
10 Pack |
For in vitro diagnostic use.