missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepare Media with 20% Glycerol
Used in the long-term frozen maintenance of bacterial stock cultures
Marca: BD 297808
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
K-Tube is glass, 100 x 13mmSpecifica
ISO/DIS, 11014 | |
2 to 25°C | |
United States | |
Microbial Cultivation | |
Long-term frozen maintenance of bacterial stock cultures | |
Ctn. of 100 size K tubes |
Beige | |
Liquid | |
Trypticase Soy Broth with 20% Glycerol | |
Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Glycerol 200mL | |
100 | |
1.5 mL, 100/Pk. |