missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Reinforced Clostridial Medium
For cultivating and enumerating clostridia, other anaerobes, and other species of bacteria from foods and clinical specimens
Marca: BD 218081
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Formulation: Peptone 10g/L, Beef extract 10g/L, Yeast extract 3g/L, Dextrose 5g/L, Sodium Chloride 5g/L, Soluble Starch 1g/L, Cysteine HCl 0.5g Sodium Acetate 3g Agar 0.5g/L
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
CCAM, EP, JP, USP

Specifica
CCAM, EP, JP, USP | |
Dehydrated | |
Reinforced Clostridial Medium | |
Cultivation and enumeration of anaerobes, particularly Clostridia from foods and clinical specimens | |
500 g |
<30°C | |
Reinforced Clostridial Medium | |
Peptone 10g/L, Beef extract 10g/L, Yeast extract 3g/L, Dextrose 5g/L, Sodium Chloride 5g/L, Soluble Starch 1g/L, Cysteine HCl 0.5g Sodium Acetate 3g Agar 0.5g/L | |
Bottle |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
BD Difco™ Dehydrated Culture Media: Reinforced Clostridial Medium > Reinforced Clostridial Medium; 500g
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto