Learn More
BD PBX Calcium Assay Kit
For homogeneous measurement of intracellular calcium changes
Marca: BD 640177
4075.63 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Includes: Calcium Indicator, 10X PBX Signal Enhancer
Descrizione
- Involves one dye addition step
- Does not require washing
- Allows easy implementation in highthroughput environment
- Probenecid, a standard inhibitor of nonspecific anion transport, is not required for dye-loading
- Can improve retention of the dye in cells
Specifica
Calcium indicator at -20°C and PBX Signal Enhancer 10X at room temperature | |
10 vials calcium indicator,100mL PBX signal enhancer (10X, lyophilized) | |
Calcium Assay Kit |
For homogeneous measurement of intracellular calcium changes | |
100 Preps |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research use only
Spot an opportunity for improvement?Share a Content Correction