Learn More
behr Labor-Technik™ behropur™ Water Deionizer Made Of Nylon
Made from Nylon. Convenient and safe mixed-bed ion exchanger for small to medium-sized quantities of high purity water. To supply laboratory glassware washers, for general laboratory maintenance and for small industrial consumers. For subsequent demineralization of reverse osmosis systems.
Marca:  behr Labor-Technik™ B00223478
 vedi altre versioni di questo prodotto
Codice prodotto. 10136541
  • 2240.00€ / Pezzo
Spedizione stimata: 21-11-2024
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Optimum exploitation of the ion exchanger capacity due to absolute uniform water distribution in the unit
    • Can be connected to the raw water network directly and without a pressure reducer
      • Resistant to alternating pressure
        • Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
          • Also available with shutoff when the limited value is reached and limit control
            • Pressure resistant to 8 bar
Specifica
Specifica
B22DNZ Pressure resistant mixed-bed unit made of nylon, permanent pressure resistant to 8 bar, spare cartridge
500 L/h
B22dNZ
2400 L
Nylon
Demineralizers