Learn More
behr Labor-Technik™ behropur™ Water Deionizer Made Of Nylon
Made from Nylon. Convenient and safe mixed-bed ion exchanger for small to medium-sized quantities of high purity water. To supply laboratory glassware washers, for general laboratory maintenance and for small industrial consumers. For subsequent demineralization of reverse osmosis systems.
Marca: behr Labor-Technik™ B00223480
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Optimum exploitation of the ion exchanger capacity due to absolute uniform water distribution in the unit
- Can be connected to the raw water network directly and without a pressure reducer
- Resistant to alternating pressure
- Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
- Also available with shutoff when the limited value is reached and limit control
- Pressure resistant to 8 bar
- Pressure resistant to 8 bar
- Also available with shutoff when the limited value is reached and limit control
- Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
- Resistant to alternating pressure
- Can be connected to the raw water network directly and without a pressure reducer
Specifica
B45DNZ Pressure resistant mixed-bed unit made of nylon, permanent pressure resistant to 8 bar, spare cartridge | |
800 L/h | |
B45dNZ |
5500 L | |
Nylon | |
Demineralizers |