Learn More
Hanna Instruments™ Bench Top pH Meters
Featuring a built-in 500 rpm magnetic stirrer, extended pH range, dual-level LCD with icons for stability and buffer recognition, and temperature display in either Celsius or Fahrenheit; this pH benchtop meter is ideal for educational purposes.
Marca: Hanna Instruments™ HI20702
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
A low cost, space efficient bench top pH meter, with a built-in magnetic stirrer. The HI-208 also features automatic pH calibration, automatic temperature compensation and is supplied complete with pH electrode which incorporates the temperature sensor. Powered by mains electricity or the supplied 9V battery (run time up to 500hr).- Compact bench pH meter
- Built-in magnetic stirrer
- Mains or batteries operated
- Built-in sample beaker and electrode holder
- Dual level LCD display for pH and temperature
- pH electrode with temperature sensor included
- Automatic pH calibration and temperature compensation
Specifica
Misuratore di pH da banco | |
±0.02 | |
Automatic | |
0.01 | |
Da –5.0 a 105.0 °C | |
LCD | |
HI207 | |
Da –2 a 16.00 °C | |
Da –5.0 a 105.0 °C | |
500 ore di utilizzo continuo |
±0.5 °C | |
Per la formazione | |
Da –2 a 16.00 °C | |
1 o 2 punti automatizzati con 2 set di tamponi memorizzati | |
0.1 °C | |
Sì | |
12 V CC | |
Automatico da –5.0 a 105.0 °C | |
Batteria da 9 V | |
203 mm |