Learn More
Hanna Instruments™ Bench Top pH Meters
Featuring a built-in 500 rpm magnetic stirrer, extended pH range, dual-level LCD with icons for stability and buffer recognition, and temperature display in either Celsius or Fahrenheit; this pH benchtop meter is ideal for educational purposes.
Marca: Hanna Instruments™ HI20802
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- A low cost, space efficient bench top pH meter, with a built-in magnetic stirrer
- Features automatic pH calibration, automatic temperature compensation and is supplied complete with pH electrode which incorporates the temperature sensor
- Designed for a busy classroom environment or can be taken outside the classroom for field studies
- Powered by mains electricity or the supplied 9V battery (run time up to 500hr)
- Compact bench pH meter
- Built-in sample beaker and electrode holder
- Dual level LCD display for pH and temperature
- pH electrode with temperature sensor included
- Automatic pH calibration and temperature compensation
Especificaciones
±0.01 | |
Da –5.0 a 105.0 °C | |
±0.01 | |
230 V | |
Automatico da –5.0 a 105.0 °C | |
Batteria da 9 V | |
190 mm | |
105 mm | |
230 V |
Da –2 a 16.00 | |
BNC | |
HI20802 | |
Da –2 a 16.00 | |
Da –5.0 a 105.0 °C | |
Misuratore di pH da banco | |
50 mm | |
230 V |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido