Learn More
Brady™ Vial Top Labels
Durable Freezerbondz™ Polyester (B-492) Vial Top Labels are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Marca: Brady™ 142919
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
- Matte white polyester material has a permanent acrylic adhesive and is thermal transfer printable
- Good print smudge resistance, solvent resistance and high/low temperature performance of -320 to 212° F (-196 to 100° C)
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Specifica
B33-144-492 | |
Acrylic, Permanent | |
Matte | |
Laboratory Label | |
Use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides | |
15.24 mm | |
1500/Pk. | |
-196°C to +130 °C | |
0.081 mm | |
0.5 x 0.5 in. | |
12.70 mm | |
12.70 mm | |
1.27 cm |
FreezerBondz Labels for the BBP33 Printer | |
White | |
Polyester | |
Non-sterile | |
BBP™33 | |
3 | |
LabelMark, MarkWare | |
0.0032 in. | |
15.88 mm | |
0.5 in. | |
0.5 in. | |
0.5 in. |