Learn More
uvex C300 Wet Cut Protection Gloves
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
8
10
Descrizione
- Soft-grip coating ensures mechanical abrasion resistance in wet applications
- Bamboo TwinFlex™ Technology provides a high level of cut resistance
- Highly flexible with good tactile sensitivity
- Silicone-free according to the imprint test
Specifica
Specifica
Area rivestita | Palm and fingertips |
Coating Material | HPE |
Colore | Green, Black |
Cut Resistance Rating | ANSI A3 |
Hand | Ambidestri |
Latex Free | Sì |
Materiale | HPPE, Polyamide, Bamboo Viscose |
Product Type | Cut-resistant Gloves |
Size | 8 |
EN388 Standard | 4X42C |
Vedi altri risultati |
Offerte speciali
-
Diversi produttori
Gear Up for Safety
Save up to 60% on gloves, apparel, and moreCodice sconto: 24131Scade: 2025-06-08
nuova offerta Shop Now
Titolo del prodotto
uvex C300 Wet Cut Protection Gloves >