Learn More
Thermo Scientific™ Capitol Vial Split Specimen Urine Collection Kit
Perform sample collection and storage with these capitol vial split foil top specimen collection kits.
Marca: Thermo Scientific™ 92023
197.00 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Descrizione
The Capitol Vial Split Specimen Collection Kit complies with SAMHSA and Department of Transportation guidelines for urine drug testing programs and other applications where a second sample is required.
Features:
- Easy-to-use kit—simply peel away the foil top lid, remove the components and provide donor with the collection cup
- After sample collection, pour 30mL of urine into the first transport vial and 15mL into the second vial, close the hinged lid until it snaps
- Place vial in the front pocket of specimen bag and completed requisition in the rear pocket
- Remove the blue release liner on the bag and fold flap over the pockets to seal
Ordering Information:
Custom branding available for foil tops and specimen bags.
Includes:
Beaker (8 oz.) with temperature indicator strip, two (2) 45mL custody vials, generic dual-pocket specimen bag, absorbent foil top.
Note:
For testing according to Substance Abuse and Mental Health Services Administration guidelines, refer to SAMSHA instruction for collection and transport requirements.
Specifica
Specimen Collection Kit | |
Non-sterile | |
U.S. Federal Government Worplace Drug Testing Programs | |
100/Cs. |
Polypropylene | |
Split Foil Top Specimen Collection Kit, 100/Cs | |
8 oz. beaker with temperature indicator strip, 2 x 45 mL Transport Tubes, a generic dual pocket specimen bag with absorbent and a foil top seal. |