Learn More
Cytiva Capto adhere
A multimodal anion exchange resin designed for post-protein A purification of monoclonal antibodies at process scale. It can also be used in bind/elute mode.
Marca:  Cytiva 17544401
Codice prodotto. 15237556
  • 807.00€ / 100mL
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Capto adhere is a multimodal BioProcess resin designed for post-protein A purification of monoclonal antibodies (MAbs) at process scale.
  • Composed of a rigid, high-flow agarose matrix and a novel multimodal ligand.
  • The strong multimodal ligand gives a different selectivity compared to traditional ion exchangers. Capto adhere can remove key contaminants such as DNA, host cell proteins (HCP), leached protein A, dimers and larger aggregates, and viruses in a single step.
  • Together with a Protein A capture step, like MabSelect SuRe, Capto adhere allows the design of a two-step chromatographic process for Mabs.
  • For optimal performance of Capto adhere, screening/optimization of loading conditions is recommended, preferably using modern high-throughput process development tools, such as PreDictor 96-well filter plates.
  • Capto adhere is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
  • As member of the BioProcess resins range, Capto adhere meets industrial demands with security of supply and comprehensive technical and regulatory support.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
100 mL
Agarose matrix
Multimodal
Ion Exchange Chromatography Media
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
Sicurezza e movimentazione
Vedi altri risultati
SDS
Documenti
Documenti
One moment while we fetch your results.
Titolo del prodotto
Cytiva Capto adhere > 100 mL