Descrizione
- Supplied without rod
- Supplied with thumbscrew
- Mounting rod hole is dual purpose and will accept both plain rods and M10 screwed rods of 12.7mm in diameter
Specifica
Ghisa | |
Supporto treppiede | |
Raccordi per aste o provette | |
2.5 kg |
Titolo del prodotto
R&L Enterprises™ Treppiedi in ghisa > Weight: 2.5kg