Learn More
Motic™ CCIS™ Plan Achromatic Strain-free Objectives
Plan Achromatic strain-free objectives for use with TEC POL Microscopes.
Marca: Motic™ 1101001707221
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- For use with Motic™ Panthera TEC POL Microscopes
- UC PL P Plan Achromatic strain-free objectives
Specifica
Microscope Objective | |
Plan Achromatic | |
0.65 |
Motic™ TEC POL Microscopes | |
40X | |
0.6 mm |