missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Antibody (Hermes-1) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 4 publications
Marca: Novus Biologicals NBP2-22530-50ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD44 Monoclonal specifically detects CD44 in Human, Mouse, Bovine, Goat, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready, Block/Neutralize.
Specifica
CD44 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:50, CyTOF-ready, Neutralization | |
CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This antibody reacts with Ovine. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF, Neutralization | |
Hermes-1 | |
Unconjugated | |
P16070 | |
CD44 | |
80 - 95 kDa lymphocyte surface glycoprotein H-CAM (CD44). | |
50 ug | |
Cancer, Cell Biology, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
960 | |
Human, Mouse, Bovine, Goat, Rabbit, Sheep | |
Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
CD44 Antibody (Hermes-1) - Azide and BSA Free, Novus Biologicals™ > 50 μg, Unlabeled
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto