Learn More
CD44 Antibody (Hermes-1) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 4 publications
Marca: Novus Biologicals NBP2-22530
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD44 Monoclonal specifically detects CD44 in Human, Mouse, Bovine, Goat, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready, Block/Neutralize.
Specifica
Monoclonal | |
CD44 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1:1 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:50, CyTOF-ready, Neutralization | |
Protein G purified |
Titolo del prodotto
CD44 Antibody (Hermes-1) - Azide and BSA Free, Novus Biologicals™ > 0.2mg; Unlabeled