Learn More
DiscoverX ChemiScreen™ GPR68/OGR1 Proton-Sensing Receptor Stable Cell Line
The ChemiScreen™ GPR68/OGR1 Proton-Sensing Receptor Stable Cell Line measures GPR68 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX HTS153C
Descrizione
The ChemiScreen™ GPR68/OGR1 Proton-Sensing Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of pH based activation of GPR68 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GPR68 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of GPR68 , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
Calcium Flux | |
Cell Lines | |
2 Vials | |
NM_003485 |
Chem-1 | |
CHECK | |
GPR68 |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.