Learn More
Metro™ Chrome Steel Shelves
Marca: Metro™ 5A357C
Dettagli aggiuntivi : Peso : 35.38000kg
Specifica
1220 x 455 mm | |
Complete Stationary Unit 5 | |
189.5 cm | |
455 mm | |
455 mm | |
Chrome Steel |
Shelf | |
For storage in ventilated area, the air flow is not disturbed | |
455 mm | |
270 kg | |
122 cm |