missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech CoraLite™488 TUNEL Assay Apoptosis Detection Kit (green)
A distinctive feature of apoptosis is degradation of DNA.
Marca: Proteintech PF00006-20T
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- The DNA fragments tend to be vary in length in integer multiples of 180 bp-200 bp, which is shown as a ladder-like pattern in agarose gel electrophoresis.
- This kit uses the TUNEL method, using Terminal Deoxynucleotidyl Transferase (TdT) to catalyze the incorporation of CL488-dUTP at the 3'-OH end of the broken DNA of apoptotic cells.
- CL488-dUTP-labeled DNA can be directly observed with a fluorescence microscope or quantified with a flow cytometer.
- The TUNEL method can selectively detect apoptotic cells, but not necrotic cells or cells with DNA strand breaks caused by irradiation and drug treatment.
Specifica
Emission: 515 nm | |
20 Tests | |
Flow Cytometry | |
Store at -20°C. Avoid exposure to light. Stable for 2 years after shipment. Avoid freeze- thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Proteintech CoraLite™488 TUNEL Assay Apoptosis Detection Kit (green) > 20 Tests
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto