Learn More
Corning™ Puntali filtranti per pipette Isotip™
Ideal for applications where avoiding cross-contamination is critical, such as DNA amplification and radioisotope handling
Marca: Corning™ 4809
Dettagli aggiuntivi : Peso : 2.16900kg
Descrizione
- IsoTip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolized contaminants from coming in contact with pipettor shafts. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website.)
- Ideal for applications where avoiding cross contamination is critical, such as DNA amplification and radioisotope handling.
- Packaged sterile.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifica
Da 100 a 1000 μl | |
Sì | |
Rack | |
Rack | |
Yes | |
Polipropilene |
Filtered | |
Incolore | |
Filtro | |
Universale | |
Isotip | |
250, 500 e 1000 μl |