missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COS-7 (African green monkey kidney) Whole cell lysate, Non-denatured; Abnova
Marca: Abnova L054V2.200ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Tissue: Kidney; Sv40 transformed
- Host: African Green Monkey
- Lysis buffer: Modified RIPA lysis buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
- Storage buffer: In modified RIPA lysis buffer
Western Blotting, Immunoprecipitation
Specifica
Western Blot | |
Whole cell lysate (non-denatured) | |
Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). | |
200 μg | |
African Green Monkey | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Kidney, SV40-transformed | |
Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
In modified RIPA Lysis Buffer. | |
5 mg/mL |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
COS-7 (African green monkey kidney) Whole cell lysate, Non-denatured; Abnova > 200μg
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto