Learn More
Sartorius Cubis™ II High-Capacity Micro Balance, 116S
GREENER_CHOICE
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
Marca:  Sartorius MCA116S-3CEU-D QP1
30215.00 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
 vedi altre versioni di questo prodotto
Codice prodotto. 17957430
  • 31255.00€ / Pezzo
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Ultra-high resolution with up to 61 million weighing steps (digits).
  • End-to-end data integrity.
  • 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management.
  • Cleanability, part of compliance.
  • Error-free operation.
  • Individual QApp workflows, motorized auto-leveling.
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer.
  • Optional inner draft shield for best weighing performance.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
MCA
Color Touchscreen
116S Weighing Module, Pharma QP1
3.5 sec.
9.4 in.
11.9 in.
111 g
0.002 mg
MCA116S-3CEU-D QP1
Balance
240 mm
302 mm