missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Cytokeratin, High Molecular Weight Ab-3, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.
Marca: Epredia™ MS-1447-R7
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
In normal cells, Ab-3 labels squamous, ductal and other complex epithelia. It reacts with benign small-acinir lesions of the prostate and does not react with hepatocytes, pancreatic acinar cells, proximal renal tubes or endometrial glands. In tumor cells, this antibody is reactive with both squamous and ductal neoplasms and variably with those derived from simple epithelium.Host Species: Mouse
Clone: 34βE12
Isotype: IgG1
Species Reactivity: Human, Monkey, Horse and Rabbit. Others not known.
Epitope: Not determined
Immunogen: Solubilized keratin extracted from human stratum corneum
Molecular Weight: 68 kDa (Cytokeratin 1), 58 kDa (Cytokeratin 5), 56.5 kDa (Cytokeratin 10) and 50 kDa (Cytokeratin 14)
Positive Control: Squamous cell CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifica
Cytokeratin, High Molecular Weight Ab-3 | |
Monoclonal | |
Unconjugated | |
Solubilized keratin extracted from human stratum corneum | |
IVD | |
Primary | |
Prediluted |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
34BetaE12 | |
Mouse | |
7 mL | |
Cytoskeletal Protein | |
Equine, Human, Non-human Primate, Rabbit | |
IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto