Learn More
Ansa di inoculazione monouso
Manufactured from polystyrene for strength and flexibility. Microspec™ Disposable Inoculation Loops are ideal for the cultivation of microbes on plates by transferring inoculum for streaking and to transfer microscopic organisms.
Marca: Microspec C3
Descrizione
- High impact, polystyrene, disposable inoculating loops
- 200mm long to reduce the risk of contamination
- Includes 50 grip seal bags of 20 ea.
Specifica
Sì | |
200 mm | |
Sterile | |
Per uso in laboratorio e ricerca | |
Laboratory and Research use |
1μL | |
Plastica | |
Inoculating Loop | |
Confezione da 1000 (50 sacchetti da 20) |
Titolo del prodotto
Ansa di inoculazione monouso > Volume: 1uL