missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Human IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Marca: Bio-Techne NBP1-73738
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifica
IgG (H+L) | |
Polyclonal | |
HRP | |
Donkey | |
Affinity Purified | |
RUO | |
Secondary | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
LYOPH | |
Western Blot 1:1000 - 1:5000, ELISA 1:10000 - 1:100000, Immunohistochemistry 1:500 - 1:2500 | |
Donkey anti-Human IgG (H+L) Secondary Antibody [HRP] (Pre-adsorbed) was produced by repeated immunization with Human IgG whole molecule in donkey. | |
1 mg | |
Adaptive Immunity, Immunology | |
This antibody was pre-adsorbed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Mouse, Rabbit, Rat, and Sheep Serum Proteins. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Mouse, Rabbit, Rat and Sheep Serum Proteins. | |
Human | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Donkey anti-Human IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™ > 1mg; HRP
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto