Learn More
Donkey anti-Mouse IgG (H+L) Secondary Antibody, Janelia Fluor 646 (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Marca: Novus Biologicals NB120-7056JF646
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Specifica
IgG (H+L) | |
Polyclonal | |
50mM Sodium Borate | |
Donkey anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed) was produced by repeated immunization with Mouse IgG whole molecule in donkey. | |
0.5 mL | |
Adaptive Immunity, Immunology | |
Mouse | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
Janelia Fluor 646 | |
Donkey | |
Multi-step | |
RUO | |
Secondary | |
Store at 4°C in the dark. |
Titolo del prodotto
Donkey anti-Mouse IgG (H+L) Secondary Antibody, Janelia Fluor 646 (Pre-adsorbed), Novus Biologicals™ > 0.5 mL; Janelia Fluor 646