Learn More
Donkey Anti-Mouse IgG NorthernLights™ NL637-conjugated Antibody, R&D Systems™
Donkey Polyclonal Antibody has been used in 9 publications
Marca: R&D Systems NL008
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG Polyclonal specifically detects IgG in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry.
Specifica
IgG | |
Polyclonal | |
NorthernLights 637 (NL637) | |
Immunoglobulin G, ImmunoglobulinG | |
Donkey | |
0.5 mL | |
Secondary | |
Mouse | |
IgG |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
1mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
IGHG1 | |
Affinity Purified | |
RUO | |
3500 | |
Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light. |
Titolo del prodotto
Donkey Anti-Mouse IgG NorthernLights™ NL637-conjugated Antibody, R&D Systems™ > 0.5mL, NL637