Learn More
Emma Safety Footwear Donovan S3 Shoe
If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Donovan.
Marca: Emma Safety Footwear MM99456947
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- S3-Safetyshoe with sole plate for wet work
- Metal Free
- The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Donovan.
- A no compromise metal free safety shoe that looks and feels like a classic sneaker.
- It gets its name from the highly supportive reinforced heel. This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
- DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
- Industries:
- Logistics & transport
- Electronics
- Construction & technology
- Manufacturing
- Nature & environment
Specifica
ISO 20345:2011 | |
Brown | |
Hydro-Tec Antibacterial | |
47 | |
High |
Laces | |
Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment | |
Nubuck, Oiled Nubuck | |
Extra-wide Width | |
Safety Shoe |