Learn More
Dräger™ Individual Haz Mat Simultaneous Test Sets
For simultaneous testing of five different gases
Marca: Dräger™ 8101736
Dettagli aggiuntivi : Peso : 0.07920kg
Allerta:
No other type of tube should be substituted with these kits.
Requires : Require adapter and a Dräger pump (sold separately) for the semi-quantitative determination of inorganic gases and organic vapors.
Descrizione
Test Set I. Includes detector tubes for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes.
Test Set II. Includes detector tubes for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene.
Test Set III. Includes detector tubes for alcohols, aliphatic hydrocarbons, aromatics, chlorinated hydrocarbons, and ketones.
Order Info
Packs of 10 test sets.
Specifica
Simultaneous Test Set II | |
For simultaneous testing of five different gases | |
Tests for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene |