Learn More
Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone
Excellent for protecting reactions against moisture while equalizing pressure. Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
Marca: Lenz Laborglasinstrumente™ 05337002
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Tube bent at a 75° angle
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifica
Bent | |
Tubo di essiccazione | |
NS 19/26 |