Learn More
DWK Life Sciences DURAN™ Screw Thread Coupling, from PBT
The screw thread coupling is used to connect two threaded connections.
Marca: DWK Life Sciences 292261055
50.16 EUR valido fino al 2025-01-31
Usa il codice promo "23894" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The screw thread coupling consists of two threaded caps, which are connected to each other via a flexible hose piece. This component enables a flexible connection of two glass components.
- For flexible connection of two glass screw-thread connections with integral silicone seal (VQM)
Specifica
Autoclavable | |
Connection of glass threads | |
GL 45 |
PBT | |
1 Ea. |