Learn More
Labconco™ Exhaust Transition Adapters for XPert™and Protector™XVS
Exhaust duct transition adapters for ductwork on Labconco Enclosures.
Marca: Labconco™ 3912400
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Exhaust duct transition adapters for ductwork on Labconco Enclosures.
Upper Exhaust Transition Adapter made of one-piece molded polyethylene. Connects 2 in. x 10 in. rectangular to 5 in. diameter round for connection to 5 in. ID hose (included with FilterMate Portable Exhauster). For use with Labconco XPert Enclosures, Protector XVS Ventilation Stations, Protector Demonstration Hoods, and Protector Work Stations.Specifica
24, 36, 48 in. XPert™ and Protector™ XVS Work Station | |
5 lb. | |
Upper Connection, 5 in. Hose | |
Epoxy-coated Steel |
10 in. | |
2.3 Kg | |
5 in. | |
Provide connection between the XPert™ Enclosures and Ductwork |