Learn More
ROBU VitraPOR™ Filter Funnel
Filter funnel available in a buchner shape or a conical shape.
Marca: ROBU VitraPOR™ 21551
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Highly resistant to most chemicals and temperatures up to 500°C
Specifica
500 mL | |
Borosilicate Glass (3.3) | |
Buchner Shape | |
1 |