Learn More
ROBU VitraPOR™ Filter Funnel
Filter funnel available in a buchner shape or a conical shape.
Marca: ROBU VitraPOR™ 22255
Dettagli aggiuntivi : Peso : 0.33720kg
Descrizione
- Highly resistant to most chemicals and temperatures up to 500°C
Specifica
25 mL | |
Conical Shape | |
10/Pk. |
Borosilicate Glass (3.3) | |
5 |