Learn More
3M™ First Base™ + Bump Cap
First Base™ + Bump Cap designed to provide limited head protection from bumps and minor lacerations from stationary objects. Patented flexible shell design manufactured from durable ABS plastic provides industrial bump protection to EN812.
Marca: 3M™ 7100207096
252.16 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Provides industrial bump protection to EN812.
- Flexible shell made with ABS plastic, can be removed for cleaning.
- Airflow channels for ventilation.
- Absorbent terry toweling sweatband.
- Available in different peak lengths for a variety of applications.
- Adjustable elastic strap for a personal fit.
- Optional logo customization areas.
- Available in different peak lengths for a variety of applications.
- Adjustable elastic strap for a personal fit.
- Belt clip accessory available.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Micro Peak | |
UV-Stabilized ABS | |
Gray | |
25 mm |