Learn More
Fisherbrand™ Temperature Probe Accessory for FB705 Sonicator
For monitoring temperature of sensitive samples
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.45000kg
Descrizione
- For monitoring temperature sensitive samples volumes
- Can be used to monitor volumes from 1 - 1000mL
- Temperature monitoring thermocouple bead on the end of a hermetically sealed wire
Specifica
Thermocouple Probe | |
Ideal for monitoring temperature of samples >1mL and indirect sonication methods | |
FB 705 Sonic Dismembrator™ |