Learn More
Fisherbrand™ accumet™ Probe
Simple Plug and Stir technology for accumet XL and AB series meters
Marca: Fisherbrand™ 01X478104
167.52 EUR valido fino al 2025-04-30
Usa il codice promo "24138" per avere il tuo prezzo promozionale.
Descrizione
Simple Plug and Stir technology for accumet XL and AB series meters
Provides mixing without magnetic stir bars or stir plates required.
- Saves valuable bench space
- Quiet operation
- Adjustable speed control
- No additional power required
- Multi-parameter XL benchtop models can accept up to two probes for simultaneous stirring
Order Info
Compatible with the following models: XL150, XL200, XL250, XL500, XL600, AB150, AB250, AB200. Replacement paddles available separately; see cat. no. 13620RP.
Servizi
Un anno
Specifica
Probe | |
Model XL150, 200, 250, 500, 600, AB150, 200 and 250 Multiparameter Benchtop Meter |
Suggerimenti di prodotto
I clienti che hanno visualizzato questo articolo hanno visto anche
Offerte speciali
-
Diversi produttori
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersCodice sconto: 25081Scade: 2025-12-31