Learn More
Sperian™ Occhiali di protezione in policarbonato Flex Seal
Fits a wide variety of workers and can be worn over prescription eyewear and with half-mask respirators. Sperian™ Honeywell™ V-Maxx™ Ventilated Safety Glasses have a sporty style for increased wearer acceptance.
Marca: Sperian™ 1006194
Descrizione
- 180° field of vision for distortion-free peripheral vision
- Rotating strap attachment system provides easier use with hard hats
- Compliant with splash resistance tests
- Indirect ventilation prevents overheating
- Anti-fog clear polycarbonate lenses
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
ISO 9001 / 2000 | |
Trasparente | |
Passaggi a impatto a 120 m/s | |
Confezione da 10 | |
Policarbonato |
Antiappannamento | |
Occhiali di sicurezza | |
Neoprene | |
Indiretto |