missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG (H+L) Secondary Antibody, PE/Cy7, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NBP1-72702PECY7
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Dot Blot, Fluorophore-linked immunosorbent assay
Specifica
IgG (H+L) | |
Polyclonal | |
PBS | |
Goat anti-Mouse IgG (H+L) Secondary Antibody was produced by repeated immunization with Mouse IgG whole molecule in goat. | |
500 μg | |
Adaptive Immunity, Immunology | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
PE-Cyanine7 | |
Goat | |
Immunogen affinity purified | |
RUO | |
Secondary | |
Store at 4°C in the dark. Do not freeze. | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Goat anti-Mouse IgG (H+L) Secondary Antibody, PE/Cy7, Novus Biologicals™ > 500 μg; PE-Cyanine7
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto