Learn More
Greiner Bio-One VACUETTE™ K2EDTA Blood Collection Tubes
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K2EDTA tubes contain dipotassium K2EDTA which blocks the coagulation cascade.
11.00€
Specifica
Product Type K2 EDTA Tube
Additivo K2 EDTA
Sterilità Sterile
Visualizza altre specifiche
Prodotti 1
Codice del prodotto Marca Colore chiusura Capacità (sistema metrico) Prezzo Quantità e disponibilità  
Codice del prodotto Marca Colore chiusura Capacità (sistema metrico) Prezzo Quantità e disponibilità  
11326470
Vedere i documenti
Greiner Bio-One
454023
Lavender with Black Ring 4 mL
11.00€
pezzi 50
Descrizione
Descrizione
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K2EDTA tubes contain dipotassium K2EDTA which blocks the coagulation cascade.
  • Tubes are supplied in packs of 50 pieces
  • For venous blood collection
  • Made out of virtually unbreakable PET plastic
  • Improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Primarily in hematology
  • Inner wall is coated with spray-dried K2EDTA which acts as an anticoagulant, binding calcium ions and interrupting the clotting cascade
  • K2EDTA Blood Collection Tubes with Gel Separator and K3EDTA Blood Collection Tubes are available
  • Pink caps for immunohematology are also available
  • CE marked
Specifica
Specifica
K2 EDTA Tube
K2 EDTA