Learn More
Greiner Bio-One VACUETTE™ K2EDTA Blood Collection Tubes
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K2EDTA tubes contain dipotassium K2EDTA which blocks the coagulation cascade.
11.00€
Codice del prodotto | Marca | Colore chiusura | Capacità (sistema metrico) | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Colore chiusura | Capacità (sistema metrico) | Prezzo | Quantità e disponibilità | ||||
11326470
|
Greiner Bio-One
454023 |
Lavender with Black Ring | 4 mL |
11.00€
pezzi 50 |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K2EDTA tubes contain dipotassium K2EDTA which blocks the coagulation cascade.
- Tubes are supplied in packs of 50 pieces
- For venous blood collection
- Made out of virtually unbreakable PET plastic
- Improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
- Primarily in hematology
- Inner wall is coated with spray-dried K2EDTA which acts as an anticoagulant, binding calcium ions and interrupting the clotting cascade
- K2EDTA Blood Collection Tubes with Gel Separator and K3EDTA Blood Collection Tubes are available
- Pink caps for immunohematology are also available
- CE marked
Specifica
K2 EDTA Tube | |
K2 EDTA | |
Sterile |