missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnium 1,1,1-trifluoro-2,4-pentanedionate, Thermo Scientific™
12.26€ - 160.00€
Identificatori chimici
CAS | 17475-68-2 |
---|---|
Formula molecolare | C20H16F12HfO8 |
Molecular Weight (g/mol) | 790.81 |
Numero MDL | MFCD00078027 |
InChI Key | JTUDTOWCQOQZBH-UHFFFAOYSA-N |
Sinonimo | Hafnium trifluoroacetylacetonate |
IUPAC Name | hafnium(4+) tetrakis(1,1,1-trifluoro-2,4-dioxopentan-3-ide) |
SMILES | [Hf+4].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11393517
|
Thermo Scientific Alfa Aesar
019275.02 |
0.5 g | N/A | ||||||
11303527
|
Thermo Scientific Alfa Aesar
019275.04 |
2 g | N/A | ||||||
11383517
|
Thermo Scientific Alfa Aesar
019275.09 |
10 g | N/A | ||||||
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificatori chimici
17475-68-2 | |
790.81 | |
JTUDTOWCQOQZBH-UHFFFAOYSA-N | |
hafnium(4+) tetrakis(1,1,1-trifluoro-2,4-dioxopentan-3-ide) |
C20H16F12HfO8 | |
MFCD00078027 | |
Hafnium trifluoroacetylacetonate | |
[Hf+4].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F |
Specifica
Hafnium 1,1,1-trifluoro-2,4-pentanedionate | |
128°C to 129°C | |
MFCD00078027 | |
Soluble in acetone, cyclohexane | |
[Hf+4].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F | |
790.81 | |
Powder |
17475-68-2 | |
C20H16F12HfO8 | |
Hafnium trifluoroacetylacetonate | |
JTUDTOWCQOQZBH-UHFFFAOYSA-N | |
hafnium(4+) tetrakis(1,1,1-trifluoro-2,4-dioxopentan-3-ide) | |
790.81 |
Sicurezza e movimentazione
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures