Learn More
Gestigkeit Harry Heating Unit with Relays
Series SGR hotplates - each heating position cam be individually and variably regulated.
Brand: Gestigkeit Harry SGR3
Additional Details : Peso : 0.10000kg
Description
- Hotplates are made of smoothly finished cast-iron with central expansion recesses. All parts of the housing are constructed from high-grade, stainless steel. Each unit has a screw-in fitting for a supporting retort rod. Designed for continuous operation.
- Regulation of Model SGR
- Each unit has an additional electronic relay and a 3-pole diode output bushing (extra-low voltage) for the contact thermometer (however this is not conditional). If used the regulation via contact thermometer takes automatic precedence. Otherwise model SGR functions as model SG, with variable regulation
Specifications
Hotplate | |
230 V | |
7.9 kg | |
50/60 Hz |
530 x 210 x 160 mm | |
500°C | |
1500 W | |
Electronic Relay |