Learn More
Scotch-Brite™ High Performance Microfibre Wipes 2010
High performance microfibre cloth that can be used on all surfaces, even delicate ones.
Marca: Scotch-Brite™ 7000103434
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- High performance microfibre wipe offering highly efficient, and lint free cleaning on virtually any surface.
- High quality cleaning results even when used with or without water, and with or without detergent
- Extremely durable, and can resist up to 500 wash cycles
- Machine washable up to 95°C
Specifica
Red | |
320 x 360 mm | |
10/Pk. |