Learn More
HMGB1/HMG-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 22 publications
Marca: Novus Biologicals NB100-2322
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated.
Specifica
| Immunohistochemistry, Immunocytochemistry | |
| Rabbit | |
| RUO |
| HMGB1 | |
| Affinity Purified | |
| Human, Rat, Canine |
Titolo del prodotto
For Research Use Only